Learn More
Surgical Design™ Pathology Tissue Retrieval Biopsy Forceps
Catalog No. 21124663 Shop All Surgical Design Products
Change view
Click to view available options
Color:
Silver, Gold
Catalog No. Color
21-124-663 Silver, Gold
1 options
Catalog No. 21-124-663 Supplier Surgical Design™ Supplier No. ATPM041
Add to Cart
Add to Cart
Angled Alexander biopsy forceps provide deep reach and controlled grip for clean tissue sampling and specimen collection during postmortem autopsy procedures.
  • Angled jaw design allows precise access to deep or recessed anatomical structures
  • Ideal for postmortem tissue sampling, including organ, tumor, mucosal, and vascular biopsies
  • Strong, serrated gripping surfaces ensure secure capture of tissue specimens
  • 11-in. length provides extended reach for large-body cavity work
  • Rigid, heavy-duty construction suitable for repeated autopsy use
  • Precision-aligned jaws for consistent tissue punch and extraction
  • Crafted from stainless steel for corrosion resistance and durability
  • Smooth, easy-to-clean surfaces optimized for pathology and morgue environments
  • Fully autoclavable and reusable
  • Commonly used by forensic pathologists, coroners, and anatomical pathology staff for consistent sampling quality
Specifications
Product Type Tissue Sampler
Autoclavable Autoclavable
Certifications/Compliance ISP, GMP
Color Silver, Gold
Material Stainless Steel
Disposable Reusable
Latex Free Yes
Sterility Non-sterile