Learn More
DGP INTELSIUS LLC PATHOPAK 1L
Supplier:  DGP INTELSIUS LLC PP011
Encompass
PathoPak 1L- PP011The 1L PathoPak provides a strong, durable and reusable, rigid secondary container that can be used for transporting Category A and UN3373 Category B Infectious Samples .Systems are supplied with all the required infectious material labelling and components to ensure compliance.Water absorbent capacity = 56 ml/1.9 fl. oz.Saline absorbent capacity = 50 ml/1.7 fl. oz.For larger purchases this product is shipped in in a box containing 20 Pathopak 1L units. The 20 unit box measures 23" x 16" x 10".
Catalog No. NC2380648