Learn More
DGP INTELSIUS LLC PATHOPAK 2L
Catalog No. NC2380646
Encompass
PP012Water absorbent capacity = 238 ml/8.3 fl. oz.Saline absorbent capacity = 215 ml/7.5 fl. oz.For larger purchases this product is shipped in in a box containing 10 Pathopak 2L units. The 10 unit box measures 32" x 13" x 8".
Catalog No. NC2380646 Supplier DGP INTELSIUS LLC Supplier No. PP012
PP012Water absorbent capacity = 238 ml/8.3 fl. oz.Saline absorbent capacity = 215 ml/7.5 fl. oz.For larger purchases this product is shipped in in a box containing 10 Pathopak 2L units. The 10 unit box measures 32" x 13" x 8".