Learn More
DGP INTELSIUS LLC PATHOPAK 3L
Catalog No. NC2380640
Encompass
PP013Comes with 2 x 6 Bay absorbents. Each 6 Bay can absorb the following:Water absorbent capacity = 238 ml/8.3 fl. oz.Saline absorbent capacity = 215 ml/7.5 fl. oz.For larger purchases this product is shipped in in a box containing 10 Pathopak 3L units. The 10 unit box measures 33" x 14" x 12".
Catalog No. NC2380640 Supplier DGP INTELSIUS LLC Supplier No. PP013
PP013Comes with 2 x 6 Bay absorbents. Each 6 Bay can absorb the following:Water absorbent capacity = 238 ml/8.3 fl. oz.Saline absorbent capacity = 215 ml/7.5 fl. oz.For larger purchases this product is shipped in in a box containing 10 Pathopak 3L units. The 10 unit box measures 33" x 14" x 12".