Learn More
Hach Company PathoScreen Field Test Kit
Supplier:  Hach Company 2859100
Encompass
PathoScreen Field Test Kit. Enough media for 100 presence-absence tests using 20 mL sample, or 20 MPN tests. Reusable glass vials, and foil-pillowed media, increases shelflife. No refrigeration required for media.
Catalog No. 50-199-6755