Learn More
Siemens Healthineers Pathromtin™ SL Reagent
Catalog No. 23044676 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
10 x 5 mL
Catalog No. Quantity
23-044-676 10 x 5 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-044-676 Supplier Siemens Healthineers Supplier No. 10446066
Add to Cart
Add to Cart
This item is not returnable. View return policy
Determination of the activated partial thromboplastin time (APTT) in citrated human plasma.
Pathromtin™ SL Reagent enables rapid screening for disorders of the intrinsic coagulation system and sensitively detects factors VIII and IX as well as the contact factors. In conjunction with deficient plasmas, it enables the individual factors of the intrinsic system to be quantified and permits diagnosis of hemophilia. It also can be used for monitoring therapy with unfractionated heparin.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Reagent
For Use With (Application) Activated Partial Thromboplastin Time (aPPT) Testing
Quantity 10 x 5 mL