Learn More
Electron Microscopy Sciences PathScan Enabler 5 Slide Scanner Low Magnification and Large Field of View
SDP
Catalog No. 501931183
Encompass
The PathScan Enabler 5 is a perfect compliment to any microscope imaging workstation. It allows scanning of whole mounted histological samples attached to standard 1 x 3 inch cover slipped glass slides. The PathScan Enabler 5 solves low power, large field of view, and high resolution imaging problems.
Scan area is 24.3 x 36.5 mm, resulting in stunning, incredible images up to 14,320 x 9,522 pixels! The PathScan Enabler 5 holds two (2) 1 x 3 inch glass slides and includes simple to use software for both Windows and Mac computers. The polarizing holder accepts a single geological 1 x 2 inch glass slide. The PathScan Enabler 5 is designed not to replace, but complement, a microscope and digital camera. The entire sample is scanned with an even, clear background and makes a perfect overview image that can be viewed in a variety of software programs. Image scans are equivalent to 4X microscope resolution.
Catalog No. 50-193-1183 Supplier Electron Microscopy Sciences Supplier No. 89015
The PathScan Enabler 5 is a perfect compliment to any microscope imaging workstation. It allows scanning of whole mounted histological samples attached to standard 1 x 3 inch cover slipped glass slides. The PathScan Enabler 5 solves low power, large field of view, and high resolution imaging problems.
Scan area is 24.3 x 36.5 mm, resulting in stunning, incredible images up to 14,320 x 9,522 pixels! The PathScan Enabler 5 holds two (2) 1 x 3 inch glass slides and includes simple to use software for both Windows and Mac computers. The polarizing holder accepts a single geological 1 x 2 inch glass slide. The PathScan Enabler 5 is designed not to replace, but complement, a microscope and digital camera. The entire sample is scanned with an even, clear background and makes a perfect overview image that can be viewed in a variety of software programs. Image scans are equivalent to 4X microscope resolution.