Learn More
Cell Signaling Technology PathScan™ Cleaved PARP Sandwich ELISA Kit, Human, Monkey, 50-Kit
SDP
Catalog No. 503133847
Encompass
The PathScan™ Cleaved PARP (Asp214) Sandwich ELISA Kit is a solid-phase enzyme-linked immunosorbent assay designed to detect endogenous levels of cleaved PARP (Asp214) protein. The kit utilizes a sandwich-based approach where the target protein is captured by a monoclonal antibody coated onto microwells, followed by detection with a biotinylated antibody and HRP-linked Streptavidin. Color development using TMB substrate allows for quantification proportional to the amount of cleaved PARP present in cell lysates.
  • Solid-phase sandwich ELISA format for precise protein detection
  • Validated for detection of endogenous cleaved PARP (Asp214) protein
  • Compatible with human and monkey samples
  • Includes all necessary reagents for efficient sample analysis
  • Allows for quantification of apoptosis via PARP cleavage
Catalog No. 50-313-3847 Supplier Cell Signaling Technology Supplier No. 7262V4
Add to Cart
Add to Cart
Request a Quote
The PathScan™ Cleaved PARP (Asp214) Sandwich ELISA Kit is a solid-phase enzyme-linked immunosorbent assay designed to detect endogenous levels of cleaved PARP (Asp214) protein. The kit utilizes a sandwich-based approach where the target protein is captured by a monoclonal antibody coated onto microwells, followed by detection with a biotinylated antibody and HRP-linked Streptavidin. Color development using TMB substrate allows for quantification proportional to the amount of cleaved PARP present in cell lysates.
  • Solid-phase sandwich ELISA format for precise protein detection
  • Validated for detection of endogenous cleaved PARP (Asp214) protein
  • Compatible with human and monkey samples
  • Includes all necessary reagents for efficient sample analysis
  • Allows for quantification of apoptosis via PARP cleavage