Learn More
Cell Signaling Technology PathScan Mono-Methyl-Histone H3 (Lys4) Sandwich ELISA Kit, Human Reactivity, 96 Assays
SDP
Catalog No. 503134135
Encompass
The PathScan™ Mono-Methyl-Histone H3 (Lys4) Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay (ELISA) designed to detect endogenous levels of histone H3 when mono-methylated at Lys4. This kit utilizes a total Histone H3 rabbit monoclonal antibody coated onto microwells to capture histone H3 from cell lysates, followed by a biotinylated detection antibody and HRP-linked streptavidin for colorimetric quantification proportional to the amount of mono-methylated protein present.
  • Solid phase sandwich ELISA format
  • Detects endogenous levels of histone H3 mono-methylated at Lys4
  • Utilizes biotinylated detection antibody and HRP-linked streptavidin
  • Supplied with a 96-well strip plate in a support frame for custom configurations
  • Includes cell lysis buffer and all necessary reagents for detection
  • Validated for use with human samples
Catalog No. 50-313-4135 Supplier Cell Signaling Technology Supplier No. 7123V2
Add to Cart
Add to Cart
Request a Quote
The PathScan™ Mono-Methyl-Histone H3 (Lys4) Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay (ELISA) designed to detect endogenous levels of histone H3 when mono-methylated at Lys4. This kit utilizes a total Histone H3 rabbit monoclonal antibody coated onto microwells to capture histone H3 from cell lysates, followed by a biotinylated detection antibody and HRP-linked streptavidin for colorimetric quantification proportional to the amount of mono-methylated protein present.
  • Solid phase sandwich ELISA format
  • Detects endogenous levels of histone H3 mono-methylated at Lys4
  • Utilizes biotinylated detection antibody and HRP-linked streptavidin
  • Supplied with a 96-well strip plate in a support frame for custom configurations
  • Includes cell lysis buffer and all necessary reagents for detection
  • Validated for use with human samples