Learn More
Cell Signaling Technology PathScan™ Phospho-Akt1 Chemiluminescent Sandwich ELISA Kit, 10 Kit
SDP
Catalog No. 503131774
Encompass
The PathScan™ Phospho-Akt1 (Ser473) Chemiluminescent Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay (ELISA) designed to detect endogenous levels of phospho-Akt1 (Ser473) protein. This kit utilizes a chemiluminescent readout, which typically offers a wider dynamic range and higher sensitivity compared to conventional chromogenic detection methods. It is provided in low volume microplates, allowing for increased sensitivity while using smaller sample volumes.
  • Provides a chemiluminescent readout proportional to the quantity of phospho-Akt1 (Ser473) protein
  • Detects endogenous levels of phospho-Akt1 (Ser473) in human, mouse, and rat samples
  • Offered in low volume microplate format for enhanced sensitivity with smaller samples
  • Custom antibody formulations specific to the kit
  • Validated for research use only
Catalog No. 50-313-1774 Supplier Cell Signaling Technology Supplier No. 7134V2
Add to Cart
Add to Cart
Request a Quote
The PathScan™ Phospho-Akt1 (Ser473) Chemiluminescent Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay (ELISA) designed to detect endogenous levels of phospho-Akt1 (Ser473) protein. This kit utilizes a chemiluminescent readout, which typically offers a wider dynamic range and higher sensitivity compared to conventional chromogenic detection methods. It is provided in low volume microplates, allowing for increased sensitivity while using smaller sample volumes.
  • Provides a chemiluminescent readout proportional to the quantity of phospho-Akt1 (Ser473) protein
  • Detects endogenous levels of phospho-Akt1 (Ser473) in human, mouse, and rat samples
  • Offered in low volume microplate format for enhanced sensitivity with smaller samples
  • Custom antibody formulations specific to the kit
  • Validated for research use only