Learn More
Cell Signaling Technology PathScan™ Phospho-Akt1 Chemiluminescent ELISA Kit, 50 Kit
SDP
Catalog No. 503132370
Encompass
The PathScan™ Phospho-Akt1 (Ser473) Chemiluminescent Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay designed to detect endogenous levels of phospho-Akt1 (Ser473) protein with a chemiluminescent readout. Utilizing low-volume microplates, this assay provides increased signal sensitivity and dynamic range while requiring smaller sample volumes compared to conventional chromogenic detection methods.
  • Detects phospho-Akt1 (Ser473) in human, mouse, and rat cells.
  • Features higher sensitivity and wider dynamic range than standard chromogenic ELISA.
  • Requires only 50 μL of sample or reagents per microwell.
  • Includes antibody-coated microwells for specific capture of phospho-Akt (Ser473).
  • Supplied in a 12 8-well strip format for flexible configuration.
  • Validated for research applications only.
Catalog No. 50-313-2370 Supplier Cell Signaling Technology Supplier No. 7134V4
Add to Cart
Add to Cart
Request a Quote
The PathScan™ Phospho-Akt1 (Ser473) Chemiluminescent Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay designed to detect endogenous levels of phospho-Akt1 (Ser473) protein with a chemiluminescent readout. Utilizing low-volume microplates, this assay provides increased signal sensitivity and dynamic range while requiring smaller sample volumes compared to conventional chromogenic detection methods.
  • Detects phospho-Akt1 (Ser473) in human, mouse, and rat cells.
  • Features higher sensitivity and wider dynamic range than standard chromogenic ELISA.
  • Requires only 50 μL of sample or reagents per microwell.
  • Includes antibody-coated microwells for specific capture of phospho-Akt (Ser473).
  • Supplied in a 12 8-well strip format for flexible configuration.
  • Validated for research applications only.