Learn More
Cell Signaling Technology PathScan™ Phospho-p53 (Ser15) Sandwich ELISA Kit, Human, 50 Kit
SDP
Catalog No. 503133961
Encompass
The PathScan™ Phospho-p53 (Ser15) Sandwich ELISA Kit is a solid-phase sandwich enzyme-linked immunosorbent assay designed to detect endogenous levels of Phospho-p53 (Ser15) protein. This kit utilizes microwells coated with a p53 rabbit monoclonal antibody to capture both phospho- and non-phospho-p53 proteins from cell lysates. Detection is performed using a phospho-p53 (Ser15) mouse monoclonal antibody followed by an HRP-linked anti-mouse IgG secondary antibody. The resulting colorimetric signal, generated by the addition of TMB substrate, is directly proportional to the quantity of captured phospho-p53 protein.
  • Validated for detection of endogenous Phospho-p53 (Ser15) protein
  • Sandwich ELISA format ensures high specificity
  • Includes HRP-linked detection system for colorimetric measurement
  • Species reactivity: human
  • Suitable for use with cell lysates
Catalog No. 50-313-3961 Supplier Cell Signaling Technology Supplier No. 7365V4
Add to Cart
Add to Cart
Request a Quote
The PathScan™ Phospho-p53 (Ser15) Sandwich ELISA Kit is a solid-phase sandwich enzyme-linked immunosorbent assay designed to detect endogenous levels of Phospho-p53 (Ser15) protein. This kit utilizes microwells coated with a p53 rabbit monoclonal antibody to capture both phospho- and non-phospho-p53 proteins from cell lysates. Detection is performed using a phospho-p53 (Ser15) mouse monoclonal antibody followed by an HRP-linked anti-mouse IgG secondary antibody. The resulting colorimetric signal, generated by the addition of TMB substrate, is directly proportional to the quantity of captured phospho-p53 protein.
  • Validated for detection of endogenous Phospho-p53 (Ser15) protein
  • Sandwich ELISA format ensures high specificity
  • Includes HRP-linked detection system for colorimetric measurement
  • Species reactivity: human
  • Suitable for use with cell lysates