Learn More
Cell Signaling Technology PathScan™ Phospho-SMAD2 (Ser465/467) Sandwich ELISA Kit, Human, Mouse, Mink, 20 Kit
SDP
Catalog No. 503133564
Encompass
The PathScan™ Phospho-SMAD2 (Ser465/467) Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay that detects endogenous levels of SMAD2 when phosphorylated at Ser465/467. This kit utilizes a SMAD2 mouse monoclonal antibody coated onto microwells to capture both phospho and non-phospho SMAD2 from cell lysates. A phospho-specific detection antibody and an HRP-linked secondary antibody are subsequently used to quantify phosphorylation levels via colorimetric substrate development.
  • Provides reliable detection of endogenous SMAD2 phosphorylation at Ser465/467
  • Validated for use with human, mouse, and mink samples
  • Includes all necessary components for colorimetric detection in a 96-well format
  • Designed for research use only
Catalog No. 50-313-3564 Supplier Cell Signaling Technology Supplier No. 7348V3
Add to Cart
Add to Cart
Request a Quote
The PathScan™ Phospho-SMAD2 (Ser465/467) Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay that detects endogenous levels of SMAD2 when phosphorylated at Ser465/467. This kit utilizes a SMAD2 mouse monoclonal antibody coated onto microwells to capture both phospho and non-phospho SMAD2 from cell lysates. A phospho-specific detection antibody and an HRP-linked secondary antibody are subsequently used to quantify phosphorylation levels via colorimetric substrate development.
  • Provides reliable detection of endogenous SMAD2 phosphorylation at Ser465/467
  • Validated for use with human, mouse, and mink samples
  • Includes all necessary components for colorimetric detection in a 96-well format
  • Designed for research use only