Learn More
Cell Signaling Technology PathScan™ Phospho-SMAD2 Sandwich ELISA Kit, Human, Mouse, Mink, 5 Kit
SDP
Catalog No. 503133930
Encompass
The PathScan™ Phospho-SMAD2 (Ser465/467) Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay that detects endogenous levels of SMAD2 when phosphorylated at Ser465/467. The kit utilizes a SMAD2 mouse monoclonal antibody coated onto microwells to capture SMAD2 protein from cell lysates. A phospho-specific detection antibody is subsequently used to identify serine phosphorylation, followed by an HRP-linked secondary antibody for colorimetric detection using TMB substrate.
  • Quantitative detection of SMAD2 phosphorylated at Ser465/467
  • Validated for use in human, mouse, and mink samples
  • Includes all necessary reagents for sandwich ELISA
  • Colorimetric detection method
  • Compatible with cell lysates
Catalog No. 50-313-3930 Supplier Cell Signaling Technology Supplier No. 7348V1
The PathScan™ Phospho-SMAD2 (Ser465/467) Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay that detects endogenous levels of SMAD2 when phosphorylated at Ser465/467. The kit utilizes a SMAD2 mouse monoclonal antibody coated onto microwells to capture SMAD2 protein from cell lysates. A phospho-specific detection antibody is subsequently used to identify serine phosphorylation, followed by an HRP-linked secondary antibody for colorimetric detection using TMB substrate.
  • Quantitative detection of SMAD2 phosphorylated at Ser465/467
  • Validated for use in human, mouse, and mink samples
  • Includes all necessary reagents for sandwich ELISA
  • Colorimetric detection method
  • Compatible with cell lysates