Learn More
Cell Signaling Technology PathScan™ RP Acetyl-α-Tubulin Sandwich ELISA Kit, 10 Kit
SDP
Catalog No. 503134204
Encompass
The PathScan™ RP Acetyl-α-Tubulin (Lys40) Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay designed to detect endogenous levels of α-tubulin protein acetylated at Lys40. Utilizing a rapid protocol, this kit enables detection in 1.5 hours. The assay functions by incubating cell lysates and a detection antibody on a coated microwell plate to form a sandwich in a single step, followed by signal development with TMB substrate.
  • Detects endogenous levels of acetylated α-tubulin (Lys40)
  • Reduced assay time of 1.5 hours
  • Compatible with human, mouse, and rat samples
  • Includes custom-formulated detection antibodies
  • Designed for use with cell lysates
Catalog No. 50-313-4204 Supplier Cell Signaling Technology Supplier No. 62490V2
Add to Cart
Add to Cart
Request a Quote
The PathScan™ RP Acetyl-α-Tubulin (Lys40) Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay designed to detect endogenous levels of α-tubulin protein acetylated at Lys40. Utilizing a rapid protocol, this kit enables detection in 1.5 hours. The assay functions by incubating cell lysates and a detection antibody on a coated microwell plate to form a sandwich in a single step, followed by signal development with TMB substrate.
  • Detects endogenous levels of acetylated α-tubulin (Lys40)
  • Reduced assay time of 1.5 hours
  • Compatible with human, mouse, and rat samples
  • Includes custom-formulated detection antibodies
  • Designed for use with cell lysates