Learn More
Cell Signaling Technology PathScan™ RP ApoE4 Sandwich ELISA Kit, Human, 50 Kit
SDP
Catalog No. 503134302
Encompass
The PathScan™ RP ApoE4 Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay designed for the detection of endogenous levels of human ApoE4 protein. This rapid protocol (RP) kit enables detection in 1.5 hours by utilizing a single-step incubation of cell lysates and detection antibody on the coated microwell plate. The assay magnitude is proportional to the quantity of ApoE4 present in the sample.
  • Rapid assay protocol completed in 1.5 hours
  • Validated for the detection of endogenous human ApoE4
  • Highly specific; does not cross-react with ApoE2 or ApoE3 isoforms
  • Includes strip-well plate format for flexible configuration
  • Requires dilution of included wash and lysis buffers prior to use
  • Compatible with standard spectrophotometric detection at 450 nm
Catalog No. 50-313-4302 Supplier Cell Signaling Technology Supplier No. 76145V4
Add to Cart
Add to Cart
Request a Quote
The PathScan™ RP ApoE4 Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay designed for the detection of endogenous levels of human ApoE4 protein. This rapid protocol (RP) kit enables detection in 1.5 hours by utilizing a single-step incubation of cell lysates and detection antibody on the coated microwell plate. The assay magnitude is proportional to the quantity of ApoE4 present in the sample.
  • Rapid assay protocol completed in 1.5 hours
  • Validated for the detection of endogenous human ApoE4
  • Highly specific; does not cross-react with ApoE2 or ApoE3 isoforms
  • Includes strip-well plate format for flexible configuration
  • Requires dilution of included wash and lysis buffers prior to use
  • Compatible with standard spectrophotometric detection at 450 nm