Learn More
Cell Signaling Technology PathScan™ RP Cleaved Gasdermin D Sandwich ELISA Kit, Mouse, 10 Kit
SDP
Catalog No. 503131932
Encompass
The PathScan™ RP Cleaved Gasdermin D (Asp276) Sandwich ELISA Kit is a solid-phase sandwich enzyme-linked immunosorbent assay designed to detect endogenous levels of the amino fragment of Gasdermin D protein cleaved at Asp276. This rapid protocol assay enables detection in approximately 1.5 hours by utilizing a single-step incubation of cell lysates and detection antibody on the coated microwell plate. The resulting signal development, measured via absorbance after the addition of TMB reagent, is directly proportional to the quantity of the cleaved protein present in the sample.
  • Provides rapid detection in 1.5 hours
  • Detects endogenous levels of mouse Gasdermin D protein cleaved at Asp276
  • Validated for high-sensitivity performance
  • Includes all necessary components for a 96-well sandwich ELISA format
Catalog No. 50-313-1932 Supplier Cell Signaling Technology Supplier No. 25566V2
Add to Cart
Add to Cart
Request a Quote
The PathScan™ RP Cleaved Gasdermin D (Asp276) Sandwich ELISA Kit is a solid-phase sandwich enzyme-linked immunosorbent assay designed to detect endogenous levels of the amino fragment of Gasdermin D protein cleaved at Asp276. This rapid protocol assay enables detection in approximately 1.5 hours by utilizing a single-step incubation of cell lysates and detection antibody on the coated microwell plate. The resulting signal development, measured via absorbance after the addition of TMB reagent, is directly proportional to the quantity of the cleaved protein present in the sample.
  • Provides rapid detection in 1.5 hours
  • Detects endogenous levels of mouse Gasdermin D protein cleaved at Asp276
  • Validated for high-sensitivity performance
  • Includes all necessary components for a 96-well sandwich ELISA format