Learn More
Cell Signaling Technology PathScan™ RP Sandwich ELISA Kit, Human, EGFR L858R Mutant, 20 Kit
SDP
Catalog No. 503131817
Encompass
This solid phase sandwich enzyme-linked immunosorbent assay (ELISA) kit is designed for the detection of endogenous levels of the EGFR (L858R) mutant protein in human cell lysates. Utilizing a rapid protocol, the assay can be completed in 1.5 hr by forming a sandwich with the mutant protein on the pre-coated microwell plate in a single-step incubation. The amount of mutant protein present is quantitatively determined by the magnitude of absorbance developed after the addition of TMB reagent and measurement at 450 nm.
  • Rapid assay completion time of 1.5 hr.
  • Designed for the specific detection of the EGFR (L858R) mutation.
  • Includes pre-coated microwells and necessary reagents for a streamlined workflow.
  • Quantitative analysis via spectrophotometric measurement at 450 nm.
  • Validated for use with human samples.
Catalog No. 50-313-1817 Supplier Cell Signaling Technology Supplier No. 19514V3
Add to Cart
Add to Cart
Request a Quote
This solid phase sandwich enzyme-linked immunosorbent assay (ELISA) kit is designed for the detection of endogenous levels of the EGFR (L858R) mutant protein in human cell lysates. Utilizing a rapid protocol, the assay can be completed in 1.5 hr by forming a sandwich with the mutant protein on the pre-coated microwell plate in a single-step incubation. The amount of mutant protein present is quantitatively determined by the magnitude of absorbance developed after the addition of TMB reagent and measurement at 450 nm.
  • Rapid assay completion time of 1.5 hr.
  • Designed for the specific detection of the EGFR (L858R) mutation.
  • Includes pre-coated microwells and necessary reagents for a streamlined workflow.
  • Quantitative analysis via spectrophotometric measurement at 450 nm.
  • Validated for use with human samples.