Learn More
Cell Signaling Technology PathScan™ RP GDF15/MIC1 Sandwich ELISA Kit, Human, 20 Kit
SDP
Catalog No. 503133287
Encompass
The PathScan™ RP GDF15/MIC1 Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay designed for the detection of endogenous levels of GDF15/MIC1 protein in a reduced assay time of 1.5 hours. In this one-step protocol, cell lysates and detection antibody are incubated together on the coated microwell plate to form a sandwich. Subsequent to washing, TMB reagent is applied for signal development, with the resulting absorbance being proportional to the GDF15/MIC1 protein quantity.
  • Detects endogenous levels of GDF15/MIC1 protein
  • Recognizes only pro-GDF15/MIC1 protein
  • Rapid assay time of 1.5 hours
  • Includes custom formulation antibodies specific to this kit
  • Supports custom plate configurations using 8-well strips
Catalog No. 50-313-3287 Supplier Cell Signaling Technology Supplier No. 75842V3
Add to Cart
Add to Cart
Request a Quote
The PathScan™ RP GDF15/MIC1 Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay designed for the detection of endogenous levels of GDF15/MIC1 protein in a reduced assay time of 1.5 hours. In this one-step protocol, cell lysates and detection antibody are incubated together on the coated microwell plate to form a sandwich. Subsequent to washing, TMB reagent is applied for signal development, with the resulting absorbance being proportional to the GDF15/MIC1 protein quantity.
  • Detects endogenous levels of GDF15/MIC1 protein
  • Recognizes only pro-GDF15/MIC1 protein
  • Rapid assay time of 1.5 hours
  • Includes custom formulation antibodies specific to this kit
  • Supports custom plate configurations using 8-well strips