Learn More
Cell Signaling Technology PathScan™ RP Phospho-eIF4E (Ser209) Sandwich ELISA Kit, 50 Kit
SDP
Catalog No. 503133420
Encompass
The PathScan™ RP Phospho-eIF4E (Ser209) Sandwich ELISA Kit is a solid-phase sandwich enzyme-linked immunosorbent assay (ELISA) designed to detect endogenous levels of eIF4E protein phosphorylated at Ser209. Utilizing a rapid protocol, this kit provides accurate results in an assay time of 1.5 hours. It functions by forming a sandwich between the capture antibody coated on the microwell plate, the target phosphorylated protein in the cell lysate, and the detection antibody in a single-step incubation.
  • Detection of endogenous eIF4E phosphorylated at Ser209
  • Reduced assay time of 1.5 hours
  • Solid-phase sandwich ELISA format
  • High specificity through custom antibody formulations
  • Includes necessary components for analysis, such as antibody-coated microwells and lysis buffer
Catalog No. 50-313-3420 Supplier Cell Signaling Technology Supplier No. 7938V4
Add to Cart
Add to Cart
Request a Quote
The PathScan™ RP Phospho-eIF4E (Ser209) Sandwich ELISA Kit is a solid-phase sandwich enzyme-linked immunosorbent assay (ELISA) designed to detect endogenous levels of eIF4E protein phosphorylated at Ser209. Utilizing a rapid protocol, this kit provides accurate results in an assay time of 1.5 hours. It functions by forming a sandwich between the capture antibody coated on the microwell plate, the target phosphorylated protein in the cell lysate, and the detection antibody in a single-step incubation.
  • Detection of endogenous eIF4E phosphorylated at Ser209
  • Reduced assay time of 1.5 hours
  • Solid-phase sandwich ELISA format
  • High specificity through custom antibody formulations
  • Includes necessary components for analysis, such as antibody-coated microwells and lysis buffer