Learn More
Cell Signaling Technology PathScan™ RP Phospho-FAK (Tyr397) Sandwich ELISA Kit, Human, 20 Kit
SDP
Catalog No. 503131555
Encompass
The PathScan™ RP Phospho-FAK (Tyr397) Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay designed for the detection of endogenous FAK protein phosphorylated at Tyr397. This kit utilizes a rapid protocol, enabling the detection of target protein in a reduced assay time of 1.5 hours through a single-step incubation of cell lysates and detection antibody.
  • Detects endogenous levels of FAK protein phosphorylated at Tyr397
  • Reduced assay time of 1.5 hours
  • Provides a single-step sandwich formation for efficient detection
  • Includes custom antibody formulations optimized for the assay
  • Compatible with human species
  • Kit contents scale proportionally with size
Catalog No. 50-313-1555 Supplier Cell Signaling Technology Supplier No. 55400V3
Add to Cart
Add to Cart
Request a Quote
The PathScan™ RP Phospho-FAK (Tyr397) Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay designed for the detection of endogenous FAK protein phosphorylated at Tyr397. This kit utilizes a rapid protocol, enabling the detection of target protein in a reduced assay time of 1.5 hours through a single-step incubation of cell lysates and detection antibody.
  • Detects endogenous levels of FAK protein phosphorylated at Tyr397
  • Reduced assay time of 1.5 hours
  • Provides a single-step sandwich formation for efficient detection
  • Includes custom antibody formulations optimized for the assay
  • Compatible with human species
  • Kit contents scale proportionally with size