Learn More
Cell Signaling Technology PathScan™ RP Phospho-IGF-I Receptor Beta Sandwich ELISA Kit, Human, 5 Kit
SDP
Catalog No. 503133293
Encompass
The PathScan™ RP Phospho-IGF-I Receptor beta (Tyr1131) Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay (ELISA) designed for the sensitive and rapid detection of endogenous levels of IGF-I receptor beta protein when phosphorylated at Tyr1131. The rapid protocol (RP) enables the assay to be performed in 1.5 hours by allowing the simultaneous incubation of cell lysate and detection antibody on the coated microwell plate.
  • Detection of endogenous IGF-I receptor beta protein phosphorylated at Tyr1131
  • Rapid 1.5 hour assay time
  • Solid phase sandwich ELISA format
  • Validated for human species reactivity
  • Custom formulation antibodies specific to kit
Catalog No. 50-313-3293 Supplier Cell Signaling Technology Supplier No. 7302V1
The PathScan™ RP Phospho-IGF-I Receptor beta (Tyr1131) Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay (ELISA) designed for the sensitive and rapid detection of endogenous levels of IGF-I receptor beta protein when phosphorylated at Tyr1131. The rapid protocol (RP) enables the assay to be performed in 1.5 hours by allowing the simultaneous incubation of cell lysate and detection antibody on the coated microwell plate.
  • Detection of endogenous IGF-I receptor beta protein phosphorylated at Tyr1131
  • Rapid 1.5 hour assay time
  • Solid phase sandwich ELISA format
  • Validated for human species reactivity
  • Custom formulation antibodies specific to kit