Learn More
Cell Signaling Technology PathScan™ RP Phospho-IGF-I Receptor β Sandwich ELISA Kit, Human, 960 Assays
SDP
Catalog No. 503134003
Encompass
The PathScan™ RP Phospho-IGF-I Receptor beta (Tyr1131) Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay that detects endogenous levels of IGF-I receptor beta protein when phosphorylated at Tyr1131. This rapid protocol kit completes in 1.5 hours by allowing the simultaneous incubation of cell lysate and detection antibody on the coated microwell plate. The magnitude of the developed color absorbance is directly proportional to the quantity of phosphorylated protein present.
  • Reduced assay time of 1.5 hours
  • Detects endogenous levels of phosphorylated IGF-I receptor beta (Tyr1131)
  • Solid phase sandwich ELISA format
  • Validated for human species reactivity
  • Custom formulation antibodies specific to kit
Catalog No. 50-313-4003 Supplier Cell Signaling Technology Supplier No. 7302V2
Add to Cart
Add to Cart
Request a Quote
The PathScan™ RP Phospho-IGF-I Receptor beta (Tyr1131) Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay that detects endogenous levels of IGF-I receptor beta protein when phosphorylated at Tyr1131. This rapid protocol kit completes in 1.5 hours by allowing the simultaneous incubation of cell lysate and detection antibody on the coated microwell plate. The magnitude of the developed color absorbance is directly proportional to the quantity of phosphorylated protein present.
  • Reduced assay time of 1.5 hours
  • Detects endogenous levels of phosphorylated IGF-I receptor beta (Tyr1131)
  • Solid phase sandwich ELISA format
  • Validated for human species reactivity
  • Custom formulation antibodies specific to kit