Learn More
Cell Signaling Technology PathScan™ RP Phospho-IRF-7 Sandwich ELISA Kit, Mouse, 20 Kit
SDP
Catalog No. 503133247
Encompass
The PathScan™ RP Phospho-IRF-7 (Ser437/438) Sandwich ELISA Kit is a solid-phase sandwich enzyme-linked immunosorbent assay that detects endogenous levels of mouse IRF-7 protein phosphorylated at Ser437/438. This rapid protocol (RP) kit enables detection in a reduced assay time of 1.5 hr by utilizing a single-step incubation for cell lysates and detection antibody on the coated microwell plate. The magnitude of the developed color absorbance is proportional to the quantity of the phosphorylated protein.
  • Reduced assay time of 1.5 hr
  • Detects endogenous mouse IRF-7 protein phosphorylated at Ser437/438
  • Solid-phase sandwich ELISA format
  • Includes IRF-7 rabbit monoclonal antibody coated microwells
  • Sensitive, rapid protocol for detection of phosphorylated target protein
Catalog No. 50-313-3247 Supplier Cell Signaling Technology Supplier No. 62491V3
Add to Cart
Add to Cart
Request a Quote
The PathScan™ RP Phospho-IRF-7 (Ser437/438) Sandwich ELISA Kit is a solid-phase sandwich enzyme-linked immunosorbent assay that detects endogenous levels of mouse IRF-7 protein phosphorylated at Ser437/438. This rapid protocol (RP) kit enables detection in a reduced assay time of 1.5 hr by utilizing a single-step incubation for cell lysates and detection antibody on the coated microwell plate. The magnitude of the developed color absorbance is proportional to the quantity of the phosphorylated protein.
  • Reduced assay time of 1.5 hr
  • Detects endogenous mouse IRF-7 protein phosphorylated at Ser437/438
  • Solid-phase sandwich ELISA format
  • Includes IRF-7 rabbit monoclonal antibody coated microwells
  • Sensitive, rapid protocol for detection of phosphorylated target protein