Learn More
Cell Signaling Technology PathScan™ RP Phospho-RIP (Ser166) Chemiluminescent ELISA Kit, 20 Kit
SDP
Catalog No. 503134070
Encompass
The PathScan™ RP Phospho-RIP (Ser166) Chemiluminescent Sandwich ELISA Kit is a solid-phase enzyme-linked immunosorbent assay designed to detect endogenous levels of RIP protein phosphorylated at Ser166. This kit utilizes a rapid 1.5 hr protocol by combining cell lysate incubation and detection antibody into a single step, providing increased sensitivity and signal in low-volume microplates. The light emission is measured in relative light units (RLU) and is proportional to the quantity of phosphorylated RIP present.
  • Rapid 1.5 hr assay time using a single-step incubation
  • High sensitivity and increased signal for precise detection
  • Compatible with smaller sample volumes
  • Species reactivity confirmed for human and mouse
  • Includes pre-coated microwell plates
Catalog No. 50-313-4070 Supplier Cell Signaling Technology Supplier No. 88918V3
Add to Cart
Add to Cart
Request a Quote
The PathScan™ RP Phospho-RIP (Ser166) Chemiluminescent Sandwich ELISA Kit is a solid-phase enzyme-linked immunosorbent assay designed to detect endogenous levels of RIP protein phosphorylated at Ser166. This kit utilizes a rapid 1.5 hr protocol by combining cell lysate incubation and detection antibody into a single step, providing increased sensitivity and signal in low-volume microplates. The light emission is measured in relative light units (RLU) and is proportional to the quantity of phosphorylated RIP present.
  • Rapid 1.5 hr assay time using a single-step incubation
  • High sensitivity and increased signal for precise detection
  • Compatible with smaller sample volumes
  • Species reactivity confirmed for human and mouse
  • Includes pre-coated microwell plates