Learn More
Cell Signaling Technology PathScan™ RP Total Aurora A Sandwich ELISA Kit, 96 Assays, 50 Kit
SDP
Catalog No. 503132550
Encompass
The PathScan™ RP Total Aurora A Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay that detects endogenous levels of Aurora A in a reduced assay time of 1.5 hours. The kit utilizes a single-step incubation of cell lysates and detection antibody on a coated microwell plate to form a sandwich with Aurora A, followed by washing and TMB reagent signal development. The magnitude of absorbance is proportional to the Aurora A quantity present in the sample.
  • Detection of endogenous Aurora A levels
  • Rapid 1.5 hour assay time
  • Includes 96-well strip plate in a support frame for custom configurations
  • Optimized for human species reactivity
  • Validated for research use
Catalog No. 50-313-2550 Supplier Cell Signaling Technology Supplier No. 46544V4
Add to Cart
Add to Cart
Request a Quote
The PathScan™ RP Total Aurora A Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay that detects endogenous levels of Aurora A in a reduced assay time of 1.5 hours. The kit utilizes a single-step incubation of cell lysates and detection antibody on a coated microwell plate to form a sandwich with Aurora A, followed by washing and TMB reagent signal development. The magnitude of absorbance is proportional to the Aurora A quantity present in the sample.
  • Detection of endogenous Aurora A levels
  • Rapid 1.5 hour assay time
  • Includes 96-well strip plate in a support frame for custom configurations
  • Optimized for human species reactivity
  • Validated for research use