Learn More
Cell Signaling Technology PathScan RP Total c-Fos Sandwich ELISA Kit, 10 Kit
SDP
Catalog No. 503132069
Encompass
The PathScan™ RP Total c-Fos Sandwich ELISA Kit is a solid-phase enzyme-linked immunosorbent assay (ELISA) designed to detect endogenous levels of c-Fos protein with a rapid, 1.5-hour protocol. This kit utilizes a single-step incubation process for cell lysates and detection antibody on a pre-coated microwell plate, followed by washing and TMB reagent addition for colorimetric detection proportional to the protein quantity present in the sample.
  • Reduced 1.5-hour assay time
  • Validated for human and mouse reactivity
  • Single-step incubation of samples and detection antibody
  • Twelve 8-well strips in a support frame for flexible configuration
  • Includes all necessary reagents for solid-phase sandwich ELISA
Catalog No. 50-313-2069 Supplier Cell Signaling Technology Supplier No. 70217V2
Add to Cart
Add to Cart
Request a Quote
The PathScan™ RP Total c-Fos Sandwich ELISA Kit is a solid-phase enzyme-linked immunosorbent assay (ELISA) designed to detect endogenous levels of c-Fos protein with a rapid, 1.5-hour protocol. This kit utilizes a single-step incubation process for cell lysates and detection antibody on a pre-coated microwell plate, followed by washing and TMB reagent addition for colorimetric detection proportional to the protein quantity present in the sample.
  • Reduced 1.5-hour assay time
  • Validated for human and mouse reactivity
  • Single-step incubation of samples and detection antibody
  • Twelve 8-well strips in a support frame for flexible configuration
  • Includes all necessary reagents for solid-phase sandwich ELISA