Learn More
Cell Signaling Technology PathScan™ RP Total CSF-1R/M-CSF-R Sandwich ELISA Kit, Human, 50 Kit
SDP
Catalog No. 503133076
Encompass
The PathScan™ RP Total CSF-1R/M-CSF-R Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay that detects endogenous levels of CSF-1R/M-CSF-R protein in a reduced assay time of 1.5 hours. In this assay, cell lysates and detection antibody are incubated on a coated microwell plate to form a sandwich with the target protein in a single step. Following an extensive wash, TMB reagent is added to develop a color, where the magnitude of absorbance is directly proportional to the quantity of CSF-1R/M-CSF-R present.
  • Rapid 1.5 hour assay time
  • Includes antibody coated microwells and detection antibody
  • Optimized for detection of human CSF-1R/M-CSF-R
  • Solid phase sandwich ELISA format for reliable quantification
Catalog No. 50-313-3076 Supplier Cell Signaling Technology Supplier No. 13032V4
Add to Cart
Add to Cart
Request a Quote
The PathScan™ RP Total CSF-1R/M-CSF-R Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay that detects endogenous levels of CSF-1R/M-CSF-R protein in a reduced assay time of 1.5 hours. In this assay, cell lysates and detection antibody are incubated on a coated microwell plate to form a sandwich with the target protein in a single step. Following an extensive wash, TMB reagent is added to develop a color, where the magnitude of absorbance is directly proportional to the quantity of CSF-1R/M-CSF-R present.
  • Rapid 1.5 hour assay time
  • Includes antibody coated microwells and detection antibody
  • Optimized for detection of human CSF-1R/M-CSF-R
  • Solid phase sandwich ELISA format for reliable quantification