Learn More
Cell Signaling Technology PathScan™ RP Total FGFR2 ELISA Kit, Human, Mouse, 20-Kit
SDP
Catalog No. 503131787
Encompass
The PathScan™ RP Total FGF Receptor 2 Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay that detects endogenous levels of FGF Receptor 2 protein in a reduced assay time of 1.5 hours. Incubation of cell lysates and detection antibody on the coated microwell plate forms a sandwich with the target protein in a single step, followed by TMB reagent signal development. This kit is intended for research use only.
  • Detection of endogenous protein levels in 1.5 hours.
  • Solid phase sandwich ELISA format for reliable results.
  • Includes pre-coated 8-well strip plates for flexible configuration.
  • Validated for reactivity with human and mouse samples.
  • Components optimized for streamlined experimental workflows.
Catalog No. 50-313-1787 Supplier Cell Signaling Technology Supplier No. 63440V3
Add to Cart
Add to Cart
Request a Quote
The PathScan™ RP Total FGF Receptor 2 Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay that detects endogenous levels of FGF Receptor 2 protein in a reduced assay time of 1.5 hours. Incubation of cell lysates and detection antibody on the coated microwell plate forms a sandwich with the target protein in a single step, followed by TMB reagent signal development. This kit is intended for research use only.
  • Detection of endogenous protein levels in 1.5 hours.
  • Solid phase sandwich ELISA format for reliable results.
  • Includes pre-coated 8-well strip plates for flexible configuration.
  • Validated for reactivity with human and mouse samples.
  • Components optimized for streamlined experimental workflows.