Learn More
Cell Signaling Technology PathScan™ RP Total LRRK2 Sandwich ELISA Kit, Rapid Protocol, Human and Mouse Reactivity, 20 Kit
SDP
Catalog No. 503134116
Encompass
The PathScan™ RP Total LRRK2 Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay designed to detect endogenous levels of LRRK2 in a rapid 1.5-hour assay time. This kit utilizes a one-step incubation process where cell lysates and the detection antibody are added to the antibody-coated microwell plate. Signal development is achieved by adding TMB reagent, with the resulting absorbance being proportional to the amount of LRRK2 present.
  • Provides rapid detection in 1.5 hours
  • Detects endogenous LRRK2 levels
  • Validated for human and mouse species reactivity
  • Enables custom plate configurations using a 96-well strip plate in a support frame
  • Includes all necessary reagents for a sandwich ELISA
Catalog No. 50-313-4116 Supplier Cell Signaling Technology Supplier No. 69930V3
Add to Cart
Add to Cart
Request a Quote
The PathScan™ RP Total LRRK2 Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay designed to detect endogenous levels of LRRK2 in a rapid 1.5-hour assay time. This kit utilizes a one-step incubation process where cell lysates and the detection antibody are added to the antibody-coated microwell plate. Signal development is achieved by adding TMB reagent, with the resulting absorbance being proportional to the amount of LRRK2 present.
  • Provides rapid detection in 1.5 hours
  • Detects endogenous LRRK2 levels
  • Validated for human and mouse species reactivity
  • Enables custom plate configurations using a 96-well strip plate in a support frame
  • Includes all necessary reagents for a sandwich ELISA