Learn More
Cell Signaling Technology PathScan™ RP Total PLK1 Sandwich ELISA Kit, 20 Kit
SDP
Catalog No. 503131574
Encompass
The PathScan™ RP Total PLK1 Sandwich ELISA Kit is a solid-phase sandwich enzyme-linked immunosorbent assay designed for the detection of endogenous levels of PLK1 protein. This rapid protocol kit significantly reduces assay time to 1.5 hours by allowing the incubation of cell lysates and detection antibody on the coated microwell plate in a single step. Following the initial incubation and washing, TMB reagent is used for signal development, with the resulting absorbance proportional to the quantity of PLK1 present in the sample.
  • Detects endogenous levels of PLK1 protein
  • Rapid 1.5 hour assay duration
  • Streamlined single-step incubation for detection antibody and lysate
  • Reliable quantitative signal development using TMB reagent
  • Optimized for human sample analysis
Catalog No. 50-313-1574 Supplier Cell Signaling Technology Supplier No. 46651V3
Add to Cart
Add to Cart
Request a Quote
The PathScan™ RP Total PLK1 Sandwich ELISA Kit is a solid-phase sandwich enzyme-linked immunosorbent assay designed for the detection of endogenous levels of PLK1 protein. This rapid protocol kit significantly reduces assay time to 1.5 hours by allowing the incubation of cell lysates and detection antibody on the coated microwell plate in a single step. Following the initial incubation and washing, TMB reagent is used for signal development, with the resulting absorbance proportional to the quantity of PLK1 present in the sample.
  • Detects endogenous levels of PLK1 protein
  • Rapid 1.5 hour assay duration
  • Streamlined single-step incubation for detection antibody and lysate
  • Reliable quantitative signal development using TMB reagent
  • Optimized for human sample analysis