Learn More
Cell Signaling Technology PathScan™ RP Total PSA/KLK3 Sandwich ELISA Kit, Human, 96 Assays
SDP
Catalog No. 503132747
Encompass
The PathScan™ RP Total PSA/KLK3 Sandwich ELISA Kit is a solid-phase, enzyme-linked immunosorbent assay designed to detect endogenous levels of total PSA/KLK3 in cell lysates. This rapid protocol kit utilizes a single-step sandwich formation process, enabling detection in 1.5 hours. Following the addition of cell lysate and detection antibody to the coated microwells, the assay utilizes TMB reagent for colorimetric signal development, with absorbance intensity directly proportional to the total PSA/KLK3 concentration.
  • Rapid protocol allows for an assay time of 1.5 hours
  • Detects endogenous levels of total PSA/KLK3 protein
  • Consists of twelve 8-well strips in a support frame for flexible plate configurations
  • Validated for human species reactivity
  • Solid-phase sandwich ELISA format
Catalog No. 50-313-2747 Supplier Cell Signaling Technology Supplier No. 14119V3
Add to Cart
Add to Cart
Request a Quote
The PathScan™ RP Total PSA/KLK3 Sandwich ELISA Kit is a solid-phase, enzyme-linked immunosorbent assay designed to detect endogenous levels of total PSA/KLK3 in cell lysates. This rapid protocol kit utilizes a single-step sandwich formation process, enabling detection in 1.5 hours. Following the addition of cell lysate and detection antibody to the coated microwells, the assay utilizes TMB reagent for colorimetric signal development, with absorbance intensity directly proportional to the total PSA/KLK3 concentration.
  • Rapid protocol allows for an assay time of 1.5 hours
  • Detects endogenous levels of total PSA/KLK3 protein
  • Consists of twelve 8-well strips in a support frame for flexible plate configurations
  • Validated for human species reactivity
  • Solid-phase sandwich ELISA format