Learn More
Cell Signaling Technology PathScan™ RP Total RSK1 Sandwich ELISA Kit, Human, 20/Pk.
SDP
Catalog No. 503133443
Encompass
The PathScan™ RP Total RSK1 Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay designed to detect endogenous levels of total RSK1 protein. This rapid protocol kit significantly reduces assay time to 1.5 hours by allowing the incubation of cell lysate and detection antibody on the coated microwell plate in a single step. Following incubation and extensive washing, TMB reagent is added for signal development, with the resulting absorbance proportional to the amount of RSK1 present in the sample.
  • Reduced assay time of 1.5 hours
  • Detects endogenous levels of total RSK1 protein
  • Solid phase sandwich ELISA format
  • Validated for human species reactivity
  • Supplied with custom formulated antibodies
Catalog No. 50-313-3443 Supplier Cell Signaling Technology Supplier No. 7966V3
Add to Cart
Add to Cart
Request a Quote
The PathScan™ RP Total RSK1 Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay designed to detect endogenous levels of total RSK1 protein. This rapid protocol kit significantly reduces assay time to 1.5 hours by allowing the incubation of cell lysate and detection antibody on the coated microwell plate in a single step. Following incubation and extensive washing, TMB reagent is added for signal development, with the resulting absorbance proportional to the amount of RSK1 present in the sample.
  • Reduced assay time of 1.5 hours
  • Detects endogenous levels of total RSK1 protein
  • Solid phase sandwich ELISA format
  • Validated for human species reactivity
  • Supplied with custom formulated antibodies