Learn More
Cell Signaling Technology PathScan™ Total Beta-Actin Sandwich ELISA Kit, 20 Kit
SDP
Catalog No. 503134259
Encompass
The PathScan™ Total β-Actin Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay designed to detect endogenous levels of β-actin. This kit employs a β-actin rabbit antibody coated onto microwells to capture the protein, followed by detection using a β-actin mouse monoclonal antibody and an anti-mouse IgG, HRP-linked secondary antibody. Colorimetric development using TMB substrate allows for quantitative determination of β-actin levels through absorbance measurement at 450 nm.
  • Provides reliable measurement of endogenous β-actin protein levels
  • Compatible with human, mouse, rat, hamster, and monkey samples
  • Includes all necessary components for sandwich ELISA detection
  • Allows for custom plate configurations using 8-well strips
  • For research use only; not for diagnostic procedures
Catalog No. 50-313-4259 Supplier Cell Signaling Technology Supplier No. 7880V3
Add to Cart
Add to Cart
Request a Quote
The PathScan™ Total β-Actin Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay designed to detect endogenous levels of β-actin. This kit employs a β-actin rabbit antibody coated onto microwells to capture the protein, followed by detection using a β-actin mouse monoclonal antibody and an anti-mouse IgG, HRP-linked secondary antibody. Colorimetric development using TMB substrate allows for quantitative determination of β-actin levels through absorbance measurement at 450 nm.
  • Provides reliable measurement of endogenous β-actin protein levels
  • Compatible with human, mouse, rat, hamster, and monkey samples
  • Includes all necessary components for sandwich ELISA detection
  • Allows for custom plate configurations using 8-well strips
  • For research use only; not for diagnostic procedures