Learn More
Cell Signaling Technology PathScan™ Total EpCAM Sandwich ELISA Kit, Human, 50 Kit
SDP
Catalog No. 503133951
Encompass
The PathScan™ Total EpCAM Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay that detects endogenous levels of EpCAM protein. The kit utilizes microwells coated with an EpCAM rabbit monoclonal antibody to capture proteins from cell lysates, followed by detection with an EpCAM mouse monoclonal antibody and an HRP-linked anti-mouse IgG secondary antibody. Color development is initiated by the addition of TMB substrate, with the resulting absorbance proportional to the quantity of EpCAM protein present in the sample.
  • Validated for detection of endogenous levels of human EpCAM protein.
  • Includes all necessary components for colorimetric detection.
  • Utilizes customized antibody formulations for high specificity.
  • Features flexible 8-well strip plate design for custom configurations.
Catalog No. 50-313-3951 Supplier Cell Signaling Technology Supplier No. 85626V4
Add to Cart
Add to Cart
Request a Quote
The PathScan™ Total EpCAM Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay that detects endogenous levels of EpCAM protein. The kit utilizes microwells coated with an EpCAM rabbit monoclonal antibody to capture proteins from cell lysates, followed by detection with an EpCAM mouse monoclonal antibody and an HRP-linked anti-mouse IgG secondary antibody. Color development is initiated by the addition of TMB substrate, with the resulting absorbance proportional to the quantity of EpCAM protein present in the sample.
  • Validated for detection of endogenous levels of human EpCAM protein.
  • Includes all necessary components for colorimetric detection.
  • Utilizes customized antibody formulations for high specificity.
  • Features flexible 8-well strip plate design for custom configurations.