Learn More
Cell Signaling Technology PathScan™ Total Ezh2 Sandwich ELISA Kit, Human, Monkey, 50 Kit
SDP
Catalog No. 503132473
Encompass
The PathScan™ Total Ezh2 Sandwich ELISA Kit is a solid-phase sandwich enzyme-linked immunosorbent assay developed to detect endogenous levels of total Ezh2 protein. This kit utilizes an Ezh2-specific antibody coated onto microwells to capture Ezh2 protein from cell lysates. A secondary Ezh2 mouse monoclonal detection antibody is then applied, followed by an HRP-linked anti-mouse IgG antibody. The addition of TMB substrate produces a colorimetric signal, where the optical density is directly proportional to the quantity of total Ezh2 protein present in the sample.
  • Validated for detection of endogenous total Ezh2 protein
  • Compatible with Human and Monkey species
  • Provides a colorimetric readout for quantitative analysis
  • Includes reagents for sample lysis and assay development
  • Flexible custom plate configurations available with supplied 8-well strips
Catalog No. 50-313-2473 Supplier Cell Signaling Technology Supplier No. 13278V4
Add to Cart
Add to Cart
Request a Quote
The PathScan™ Total Ezh2 Sandwich ELISA Kit is a solid-phase sandwich enzyme-linked immunosorbent assay developed to detect endogenous levels of total Ezh2 protein. This kit utilizes an Ezh2-specific antibody coated onto microwells to capture Ezh2 protein from cell lysates. A secondary Ezh2 mouse monoclonal detection antibody is then applied, followed by an HRP-linked anti-mouse IgG antibody. The addition of TMB substrate produces a colorimetric signal, where the optical density is directly proportional to the quantity of total Ezh2 protein present in the sample.
  • Validated for detection of endogenous total Ezh2 protein
  • Compatible with Human and Monkey species
  • Provides a colorimetric readout for quantitative analysis
  • Includes reagents for sample lysis and assay development
  • Flexible custom plate configurations available with supplied 8-well strips