Learn More
Cell Signaling Technology PathScan™ Total ROS1 Sandwich ELISA Kit, Human, 10 Kit
SDP
Catalog No. 503131645
Encompass
The PathScan™ Total ROS1 Sandwich ELISA Kit is a solid-phase enzyme-linked immunosorbent assay designed to detect endogenous levels of total ROS1 protein in human cells. This kit employs a mouse antibody coated onto microwells to capture ROS1 protein from cell lysates. A rabbit detection antibody is subsequently added, followed by an HRP-linked anti-rabbit IgG antibody to facilitate colorimetric detection using TMB substrate. The resulting absorbance is proportional to the quantity of ROS1 protein present in the sample.
  • Specifically detects endogenous ROS1 (phospho and nonphospho) protein
  • Validated for use with human cell samples
  • Includes all necessary components for colorimetric detection
  • Flexible strip-well plate configuration for custom assays
  • Convenient ready-to-use reagents following reconstitution
Catalog No. 50-313-1645 Supplier Cell Signaling Technology Supplier No. 7091V2
Add to Cart
Add to Cart
Request a Quote
The PathScan™ Total ROS1 Sandwich ELISA Kit is a solid-phase enzyme-linked immunosorbent assay designed to detect endogenous levels of total ROS1 protein in human cells. This kit employs a mouse antibody coated onto microwells to capture ROS1 protein from cell lysates. A rabbit detection antibody is subsequently added, followed by an HRP-linked anti-rabbit IgG antibody to facilitate colorimetric detection using TMB substrate. The resulting absorbance is proportional to the quantity of ROS1 protein present in the sample.
  • Specifically detects endogenous ROS1 (phospho and nonphospho) protein
  • Validated for use with human cell samples
  • Includes all necessary components for colorimetric detection
  • Flexible strip-well plate configuration for custom assays
  • Convenient ready-to-use reagents following reconstitution