Learn More
Cell Signaling Technology PathScan™ Total SMAD2/3 Sandwich ELISA Kit, 96 Assays
SDP
Catalog No. 503133452
Encompass
The PathScan™ Total SMAD2/3 Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay that recognizes endogenous levels of total SMAD2 and SMAD3 proteins. This quantitative colorimetric assay utilizes a SMAD2/3 mouse monoclonal antibody coated on microwells to capture both phospho and nonphospho forms from cell lysates. Detection is achieved using a SMAD2/3 rabbit detection monoclonal antibody, followed by an HRP-linked anti-rabbit IgG antibody, with TMB substrate used for color development. The magnitude of absorbance is proportional to the quantity of captured SMAD2 and SMAD3 proteins.
  • Validated for reactivity in human, mouse, rat, monkey, and mink cells.
  • Quantitative colorimetric ELISA format.
  • Includes a 96-well strip plate consisting of twelve 8-well strips in a support frame for custom configurations.
  • Suitable for detection of total SMAD2 and SMAD3 proteins.
Catalog No. 50-313-3452 Supplier Cell Signaling Technology Supplier No. 12000V2
Add to Cart
Add to Cart
Request a Quote
The PathScan™ Total SMAD2/3 Sandwich ELISA Kit is a solid phase sandwich enzyme-linked immunosorbent assay that recognizes endogenous levels of total SMAD2 and SMAD3 proteins. This quantitative colorimetric assay utilizes a SMAD2/3 mouse monoclonal antibody coated on microwells to capture both phospho and nonphospho forms from cell lysates. Detection is achieved using a SMAD2/3 rabbit detection monoclonal antibody, followed by an HRP-linked anti-rabbit IgG antibody, with TMB substrate used for color development. The magnitude of absorbance is proportional to the quantity of captured SMAD2 and SMAD3 proteins.
  • Validated for reactivity in human, mouse, rat, monkey, and mink cells.
  • Quantitative colorimetric ELISA format.
  • Includes a 96-well strip plate consisting of twelve 8-well strips in a support frame for custom configurations.
  • Suitable for detection of total SMAD2 and SMAD3 proteins.