Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Patient/Exam Room Sharps Collectors

Catalog No. 1482665A Shop All Waters Corporation Products
Change view
Click to view available options
Capacity (English):
5.4 qt.
2 gal.
3 gal.
Color:
Clear
Pearl
Red
Design:
Counterbalanced Door
Counterbalanced Side Entry
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Color Design
14-826-65A 5.4 qt. Red Counterbalanced Door
14-826-145 3 gal. Red Counterbalanced Door
14-826-142 2 gal. Pearl Counterbalanced Door
14-830-29 5.4 qt. Pearl Counterbalanced Side Entry
14-826-64B 5.4 qt. Clear Counterbalanced Side Entry
14-826-65B 5.4 qt. Clear Counterbalanced Door
14-826-144 2 gal. Red Counterbalanced Door
14-830-28 5.4 qt. Red Counterbalanced Side Entry
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 14-826-65A Supplier BD Supplier No. 305426
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Horizontal, side or vertical entry

  • Strong, durable polypropylene construction gives containers excellent impact and puncture-resistance
  • Suitable for incineration or autoclaving
  • Clear top and door provide visual indication of fill level
  • Safety transport handles keep hands away from collector body

Horizontal- and Side-Entry Collectors

  • Counterbalanced door opens automatically for single-handed disposal; stays closed to prevent overfilling

Order Info

Shipped nested; assembly of top to base required.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 4.7 L
Design Counterbalanced Door
Capacity (English) 5.4 qt.
Color Red
Disposable Single-use
For Use With (Application) Suitable for incineration or autoclaving
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.