Learn More
CoaguSense Inc™ Patient Test Strips
Catalog No. 23654012
Change view
Click to view available options
CE Marker:
Self-declared
Catalog No. CE Marker
23-654-012 Self-declared
1 options
This item is not returnable. View return policy
Catalog No. 23-654-012 Supplier CoaguSense Inc™ Supplier No. 03P5750
Add to Cart
Add to Cart
This item is not returnable. View return policy
Pack of 50 PT/INR patient test strips.
CoaguSense Inc™ PT/INR test strip kit includes 50 patient test strips, 2 high and 2 low control strips, 1 control activation solution vial, 54 sample transferred tubes with plungers, and 1 NFC card. Package insert works with PT2 meters.
Specifications
CE Marker Self-declared
Clia Complexity Waived, Moderate
Format 40% Reagent Pre-mix with Diluent, Phospholipid Blend, Buffer; ph: 7.2
Type Test Strip
Disposable Single-use