Learn More
Selleck Chemical LLC Patritumab deruxtecan
SDP
Catalog No. 503134573
Encompass_Preferred
Patritumab deruxtecan (HER3-DXd) is an antibody-drug conjugate (ADC) composed of a fully human anti-HER3 IgG1 monoclonal antibody and a cytotixic topoisomerase I inhibitor Patritumab deruxtecan shows anticancer activity
Catalog No. 50-313-4573 Supplier Selleck Chemical LLC Supplier No. D40041MG5
Patritumab deruxtecan (HER3-DXd) is an antibody-drug conjugate (ADC) composed of a fully human anti-HER3 IgG1 monoclonal antibody and a cytotixic topoisomerase I inhibitor Patritumab deruxtecan shows anticancer activity