Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Standards Kit, Patrol UPLC, UV Detection, 10 mL Volume

Catalog No. 502390626 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This standards kit is suitable for performance verification of a UV Patrol System. It includes three 10 mL ampules of standard solutions. Solution 1 contains 0.1000 mg/mL Caffeine. Solution 2 contains 4.00 μg/mL each of 2-Acetylfuran, Acetanilide, Acetophenone, Propiophenone, Butylparaben, Benzophenone, and Valerophenone. Solution 3 contains 4.000 mg/mL Caffeine.

  • Detection method: UV
  • Formulation: Liquid
  • Storage condition: Room temperature
  • System type: LC
  • Instrument: PATROL UPLC
  • Quantitative: Yes
  • Shelf life: 24 Months
  • Shipping condition: Ambient
  • Volume: 10 mL per ampule
  • Application: System performance
  • Product type: Standards
  • Units per package: 1 pk

Catalog No. 50-239-0626 Supplier Waters Corp Supplier No. 700005515
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This standards kit is suitable for performance verification of a UV Patrol System. It includes three 10 mL ampules of standard solutions. Solution 1 contains 0.1000 mg/mL Caffeine. Solution 2 contains 4.00 μg/mL each of 2-Acetylfuran, Acetanilide, Acetophenone, Propiophenone, Butylparaben, Benzophenone, and Valerophenone. Solution 3 contains 4.000 mg/mL Caffeine.

  • Detection method: UV
  • Formulation: Liquid
  • Storage condition: Room temperature
  • System type: LC
  • Instrument: PATROL UPLC
  • Quantitative: Yes
  • Shelf life: 24 Months
  • Shipping condition: Ambient
  • Volume: 10 mL per ampule
  • Application: System performance
  • Product type: Standards
  • Units per package: 1 pk

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.