Learn More
Medchemexpress LLC Patulin | 149-29-1 | 10 MG
SDP
Catalog No. 5000419375
Encompass_Preferred
Patulin (Terinin) is a mycotoxin produced by fungi, including Aspergillus, Penicillium, and Byssochlamys species. It causes chromosome breakage, mutation, teratogenic, and cytotoxic effects. Patulin induces autophagy-dependent apoptosis through the lysosomal-mitochondrial axis and causes DNA damage.
  • Purity: 99.94%
  • For research use only
  • Induces autophagy-dependent apoptosis through lysosomal-mitochondrial axis
  • Causes DNA damage
  • Increases cytochrome c protein level and leads to apoptosis induction in an autophagy-dependent manner in HepG2 cells
  • Impairs mitophagy in HepG2 cells
  • Induces DNA damage in the brain, liver, and kidneys in a dose-dependent manner in CF-1 mice
Catalog No. 50-004-19375 Supplier Medchemexpress LLC Supplier No. HYN677910MG
Patulin (Terinin) is a mycotoxin produced by fungi, including Aspergillus, Penicillium, and Byssochlamys species. It causes chromosome breakage, mutation, teratogenic, and cytotoxic effects. Patulin induces autophagy-dependent apoptosis through the lysosomal-mitochondrial axis and causes DNA damage.
  • Purity: 99.94%
  • For research use only
  • Induces autophagy-dependent apoptosis through lysosomal-mitochondrial axis
  • Causes DNA damage
  • Increases cytochrome c protein level and leads to apoptosis induction in an autophagy-dependent manner in HepG2 cells
  • Impairs mitophagy in HepG2 cells
  • Induces DNA damage in the brain, liver, and kidneys in a dose-dependent manner in CF-1 mice