Learn More
Medchemexpress LLC Patulin | 149-29-1 | 5 MG
SDP
Catalog No. 5000418216
Encompass_Preferred
Patulin is a mycotoxin produced by fungi including Aspergillus, Penicillium, and Byssochlamys species. It causes chromosome breakage, mutation, teratogenic, and cytotoxic effects. Patulin induces autophagy-dependent apoptosis through the lysosomal-mitochondrial axis and causes DNA damage. This product is for research use only. Store at -20°C, protect from light. When in solvent, store at -80°C for 6 months or -20°C for 1 month, protecting from light.
  • Induces autophagy-dependent apoptosis
  • Causes DNA damage
  • Impairs mitophagy in HepG2 cells
  • Increases active cleaved caspase-3 levels
  • Increases LC3-II levels
  • Reduces intracellular glutathione concentration in vivo
  • Increases lipid peroxidation in vivo
Catalog No. 50-004-18216 Supplier Medchemexpress LLC Supplier No. HYN67795MG
Patulin is a mycotoxin produced by fungi including Aspergillus, Penicillium, and Byssochlamys species. It causes chromosome breakage, mutation, teratogenic, and cytotoxic effects. Patulin induces autophagy-dependent apoptosis through the lysosomal-mitochondrial axis and causes DNA damage. This product is for research use only. Store at -20°C, protect from light. When in solvent, store at -80°C for 6 months or -20°C for 1 month, protecting from light.
  • Induces autophagy-dependent apoptosis
  • Causes DNA damage
  • Impairs mitophagy in HepG2 cells
  • Increases active cleaved caspase-3 levels
  • Increases LC3-II levels
  • Reduces intracellular glutathione concentration in vivo
  • Increases lipid peroxidation in vivo