Learn More
Medchemexpress LLC HY-N6779 1mg Medchemexpress, Patulin CAS:149-29-1 Purity:>98%
SDP
Catalog No. 501970469
Encompass_Preferred
Medchemexpress, HY-N6779 1mg Patulin CAS:149-29-1 Patulin (Terinin) is a mycotoxin produced by fungi including the Aspergillus, Penicillium, and Byssochlamys species, is suspected to be clastogenic, mutagenic, teratogenic and cytotoxic. Patulin (Terinin) induces autophagy-dependent apoptosis through lysosomal-mitochondrial axis, and causes DNA damage[3][4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0469 Supplier Medchemexpress LLC Supplier No. HYN67791MG
Medchemexpress, HY-N6779 1mg Patulin CAS:149-29-1 Patulin (Terinin) is a mycotoxin produced by fungi including the Aspergillus, Penicillium, and Byssochlamys species, is suspected to be clastogenic, mutagenic, teratogenic and cytotoxic. Patulin (Terinin) induces autophagy-dependent apoptosis through lysosomal-mitochondrial axis, and causes DNA damage[3][4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.