Learn More
Aviva Systems Biology Corp PATZ1 Antibody - N-terminal region (ARP39077_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39077_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PATZ1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PATZ1. Alternate names: MAZR, PATZ, RIAZ, ZBTB19, ZNF278, ZSG, dJ400N23ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56902