Learn More
Cayman Chemical Brazilein, Purity: ≥95%, CAS: 600-76-0, 10 mg
SDP
Catalog No. 502913880
Encompass_Preferred
A phytochemical with diverse biological activities; active against E. coli, P. aeruginosa, and MSSA (MICs = 32, 512, and 512 µg/ml), as well as a panel of 10 clinical isolates of MRSA (MIC50s = 16-32 µg/ml); inhibits LPS-induced NO production in J774A.1 cells at 10 µM; cytotoxic to HepG2, Hep3B, MDA-MB-231, MCF-7, A549, and Ca9-22 cancer cells (IC50s = 3.15, 3.42, 2.36, 3.96, 9.68, and 8.63 µg/ml, respectively); inhibits concanavalin A-induced proliferation of isolated mouse spleen lymphocytes from 1-10 µg/ml
Catalog No. 50-291-3880 Supplier Cayman Chemical Supplier No. 3673610 MG
A phytochemical with diverse biological activities; active against E. coli, P. aeruginosa, and MSSA (MICs = 32, 512, and 512 µg/ml), as well as a panel of 10 clinical isolates of MRSA (MIC50s = 16-32 µg/ml); inhibits LPS-induced NO production in J774A.1 cells at 10 µM; cytotoxic to HepG2, Hep3B, MDA-MB-231, MCF-7, A549, and Ca9-22 cancer cells (IC50s = 3.15, 3.42, 2.36, 3.96, 9.68, and 8.63 µg/ml, respectively); inhibits concanavalin A-induced proliferation of isolated mouse spleen lymphocytes from 1-10 µg/ml