Learn More
ATCC pSV3-neo plasmid; in E. coli
Catalog No. 502379826
Encompass
Vector that provides dominant selectable marker for resistance to antibiotic G418 in mammalian cell lines. Restriction digests of the vector gave the following sizes (kb) : EcoRI-8.6 ; HindIII-4.6, 3.1, 1.4, 0.64 ; PvuII-4.0, 2.1, 1.1, 0.74, 0.64, 0.37. Depositors: Paul Berg, Biochemistry Department Stanford University Medical Center Applications: Shuttle vector, Expression vector
Catalog No. 50-237-9826 Supplier ATCC Supplier No. 37150
Vector that provides dominant selectable marker for resistance to antibiotic G418 in mammalian cell lines. Restriction digests of the vector gave the following sizes (kb) : EcoRI-8.6 ; HindIII-4.6, 3.1, 1.4, 0.64 ; PvuII-4.0, 2.1, 1.1, 0.74, 0.64, 0.37. Depositors: Paul Berg, Biochemistry Department Stanford University Medical Center Applications: Shuttle vector, Expression vector