Learn More
Medchemexpress LLC HY-14432 5mg Medchemexpress, Pavinetant CAS:941690-55-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY144325MG
Encompass_Preferred
Medchemexpress, HY-14432 5mg Pavinetant CAS:941690-55-7 Pavinetant (MLE-4901) is a neurokinin-3 receptor (NK3R) antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1601