Learn More
MSC Bare Ground Solutions Pet Safe Ice Melt, Urea, 40 lb
Catalog No. 502816890
Encompass
Paw Pleaser is a 100% non-salt, non-toxic, and pet-safe ice and snow melter. This urea-based formulation is designed to reduce environmental impact and effectively de-ice surfaces down to 10°F, while being harmless to pets, children, and landscaping.
  • Contains no harsh chemical chlorides
  • Pet and child safe, will not cause irritation to paws
  • Safe on all surfaces including grass, plants, shrubs, bricks, and concrete
  • Biodegradable and non-corrosive, with runoff that can boost lawn growth
  • Effective to 10°F
  • Non-polluting, contains no harmful methane, glycols, or methanol
Catalog No. 50-281-6890 Supplier MSC Supplier No. 19877851
Paw Pleaser is a 100% non-salt, non-toxic, and pet-safe ice and snow melter. This urea-based formulation is designed to reduce environmental impact and effectively de-ice surfaces down to 10°F, while being harmless to pets, children, and landscaping.
  • Contains no harsh chemical chlorides
  • Pet and child safe, will not cause irritation to paws
  • Safe on all surfaces including grass, plants, shrubs, bricks, and concrete
  • Biodegradable and non-corrosive, with runoff that can boost lawn growth
  • Effective to 10°F
  • Non-polluting, contains no harmful methane, glycols, or methanol